Product Details

SNP ID
rs76008787
Assay Type
Functionally tested
NCBI dbSNP Submissions
26
Location
Chr.1:56720993 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTCCATGCTTTTTCTCTGTGTGAG[C/T]TTATGTATTCTTATATATAGCTATA
Phenotype
MIM: 600497
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C1orf168 PubMed Links

Gene Details

Gene
C1orf168
Gene Name
chromosome 1 open reading frame 168
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001004303.4 Intron NP_001004303.3
XM_005270584.3 Intron XP_005270641.1
XM_011540898.1 Intron XP_011539200.1
XM_011540899.1 Intron XP_011539201.1
XM_011540900.2 Intron XP_011539202.1
XM_011540903.2 Intron XP_011539205.1
XM_011540904.2 Intron XP_011539206.1
XM_011540905.2 Intron XP_011539207.1
XM_011540906.2 Intron XP_011539208.1
XM_017000544.1 Intron XP_016856033.1
XM_017000545.1 Intron XP_016856034.1
XM_017000546.1 Intron XP_016856035.1
XM_017000547.1 Intron XP_016856036.1
XM_017000548.1 Intron XP_016856037.1
Gene
PRKAA2
Gene Name
protein kinase AMP-activated catalytic subunit alpha 2
There are no transcripts associated with this gene.

View Full Product Details