Product Details
- SNP ID
-
rs79353574
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
2
- Location
-
Chr.1:58782574 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGAGGTTTGCGTAGACCGGCGGC[G/T]CGCTGTGCAGGCTGGCGCTGAAGCC
- Phenotype
-
MIM: 165160
- Polymorphism
- G/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
JUN
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4647014] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- JUN
- Gene Name
- Jun proto-oncogene, AP-1 transcription factor subunit
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_002228.3 |
1540 |
Missense Mutation |
GAG,GCG |
E166A |
NP_002219.1 |
- Gene
- LINC01135
- Gene Name
- long intergenic non-protein coding RNA 1135
There are no transcripts associated with this gene.
View Full Product Details