Product Details

SNP ID
rs76411783
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:63595756 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGACCCAAGAGTTGTCCTTAAGGAG[C/T]TTACAGCAGAGTGGGAATTTTAGGT
Phenotype
MIM: 171900
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PGM1 PubMed Links

Gene Details

Gene
PGM1
Gene Name
phosphoglucomutase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001172818.1 Intron NP_001166289.1
NM_001172819.1 Intron NP_001166290.1
NM_002633.2 Intron NP_002624.2

View Full Product Details