Product Details

SNP ID
rs77110341
Assay Type
Functionally tested
NCBI dbSNP Submissions
20
Location
Chr.1:33011210 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAACTTTGAGGCCAAGTGCTTACAA[C/T]TGTCCCTAGTTAAAGGGGAGCTGAT
Phenotype
MIM: 103020
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
AK2 PubMed Links

Gene Details

Gene
AK2
Gene Name
adenylate kinase 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001199199.1 Intron NP_001186128.1
NM_001319139.1 Intron NP_001306068.1
NM_001319140.1 Intron NP_001306069.1
NM_001319141.1 Intron NP_001306070.1
NM_001319142.1 Intron NP_001306071.1
NM_001319143.1 Intron NP_001306072.1
NM_001625.3 Intron NP_001616.1
NM_013411.4 Intron NP_037543.1

View Full Product Details