Product Details

SNP ID
rs79557900
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:34168436 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGGGGCTGGATCCAGGTAAGCAA[A/T]GGGAGATAGGGAGTGGGGCAGATCA
Phenotype
MIM: 608398
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
C1orf94 PubMed Links

Gene Details

Gene
C1orf94
Gene Name
chromosome 1 open reading frame 94
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001134734.1 Intron NP_001128206.1
NM_032884.4 Intron NP_116273.2
Gene
CSMD2
Gene Name
CUB and Sushi multiple domains 2
There are no transcripts associated with this gene.

View Full Product Details