Product Details

SNP ID
rs75362566
Assay Type
Functionally tested
NCBI dbSNP Submissions
15
Location
Chr.1:83866912 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGTGATGATTTAAAATAAAAAAG[A/G]TTGTTTCTTAGACATTTCTAGAAAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101927560 PubMed Links

Gene Details

Gene
LOC101927560
Gene Name
uncharacterized LOC101927560
There are no transcripts associated with this gene.

Gene
LOC101927587
Gene Name
uncharacterized LOC101927587
There are no transcripts associated with this gene.

Gene
TTLL7
Gene Name
tubulin tyrosine ligase like 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024686.4 5885 Intron NP_078962.4
XM_005271208.2 5885 UTR 3 XP_005271265.1
XM_011542159.2 5885 Intron XP_011540461.1
XM_017002347.1 5885 UTR 3 XP_016857836.1
XM_017002348.1 5885 Intron XP_016857837.1
XM_017002349.1 5885 UTR 3 XP_016857838.1

View Full Product Details