Product Details

SNP ID
rs78286921
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:84629053 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTGTCTTCCTTAATTTTCCTGTAT[A/C]AACCAGATGAAGGTGTTCACAGAGG
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
LINC01461 PubMed Links

Gene Details

Gene
LINC01461
Gene Name
long intergenic non-protein coding RNA 1461
There are no transcripts associated with this gene.

Gene
LINC01555
Gene Name
long intergenic non-protein coding RNA 1555
There are no transcripts associated with this gene.

View Full Product Details