Product Details

SNP ID
rs78063516
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:86547213 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATTAAGCTGAATAATAATGGCTTT[G/T]AAGATATTGTCATTGTTATAGATCC
Phenotype
MIM: 616857
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
CLCA4 PubMed Links

Gene Details

Gene
CLCA4
Gene Name
chloride channel accessory 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012128.3 138 Nonsense Mutation GAA,TAA E32* NP_036260.2
XM_011541015.2 138 Intron XP_011539317.1

View Full Product Details