Product Details

SNP ID
rs76870834
Assay Type
Functionally Tested
NCBI dbSNP Submissions
14
Location
Chr.1:88935961 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATCCACTATGTTATGCTGACTCTTA[C/T]AAAATGATTGTGGAAGGTGTGTTAA
Phenotype
MIM: 610656
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
KYAT3 PubMed Links

Gene Details

Gene
KYAT3
Gene Name
kynurenine aminotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008661.2 1864 UTR 3 NP_001008661.1
NM_001008662.2 1864 UTR 3 NP_001008662.1

View Full Product Details