Product Details

SNP ID
rs77285828
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:88987051 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGTATGGAGAGGGGTTAGTGCTATC[C/T]CAAATAGTCCACAAGTTTGAATAAT
Phenotype
MIM: 610656
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KYAT3 PubMed Links

Gene Details

Gene
KYAT3
Gene Name
kynurenine aminotransferase 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008661.2 Intron NP_001008661.1
NM_001008662.2 Intron NP_001008662.1
Gene
RBMXL1
Gene Name
RNA binding motif protein, X-linked like 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001162536.2 Intron NP_001156008.1
NM_019610.5 Intron NP_062556.2

View Full Product Details