Product Details

SNP ID
rs79423278
Assay Type
Functionally tested
NCBI dbSNP Submissions
10
Location
Chr.1:35558686 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTCCGGATAATCGAACTTCACCCAT[G/T]TATGTCTCCATTTCTCCCTGTCTGT
Phenotype
MIM: 613535 MIM: 608458
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
KIAA0319L PubMed Links

Gene Details

Gene
KIAA0319L
Gene Name
KIAA0319 like
There are no transcripts associated with this gene.

Gene
NCDN
Gene Name
neurochondrin
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001014839.1 Intron NP_001014839.1
NM_001014841.1 Intron NP_001014841.1
NM_014284.2 Intron NP_055099.1

View Full Product Details