Product Details

SNP ID
rs74865632
Assay Type
Functionally tested
NCBI dbSNP Submissions
5
Location
Chr.1:92510035 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAAGGCATAATATCCTATTGCATCC[C/T]AAAGGCCTGGGAGATTTTAAACTCT
Phenotype
MIM: 602942
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
EVI5 PubMed Links

Gene Details

Gene
EVI5
Gene Name
ecotropic viral integration site 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308248.1 6064 UTR 3 NP_001295177.1
NM_005665.5 6064 UTR 3 NP_005656.4
XM_017002269.1 6064 Intron XP_016857758.1
XM_017002270.1 6064 Intron XP_016857759.1
XM_017002271.1 6064 Intron XP_016857760.1
XM_017002272.1 6064 UTR 3 XP_016857761.1
XM_017002273.1 6064 Intron XP_016857762.1
XM_017002274.1 6064 Intron XP_016857763.1
XM_017002275.1 6064 Intron XP_016857764.1
XM_017002276.1 6064 Intron XP_016857765.1
XM_017002277.1 6064 Intron XP_016857766.1
XM_017002278.1 6064 Intron XP_016857767.1
XM_017002279.1 6064 Intron XP_016857768.1
XM_017002280.1 6064 Intron XP_016857769.1
XM_017002281.1 6064 Intron XP_016857770.1
XM_017002282.1 6064 Intron XP_016857771.1
XM_017002283.1 6064 Intron XP_016857772.1
XM_017002284.1 6064 Intron XP_016857773.1
XM_017002285.1 6064 Intron XP_016857774.1
XM_017002286.1 6064 Intron XP_016857775.1
XM_017002287.1 6064 Intron XP_016857776.1
XM_017002288.1 6064 Intron XP_016857777.1

View Full Product Details