Product Details

SNP ID
rs76010229
Assay Type
Functionally tested
NCBI dbSNP Submissions
9
Location
Chr.1:35927000 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAATATTGAGGGACCTTTCTAGG[A/G]AGCCTTTGGTCCTTTGTCCCCATTT
Phenotype
MIM: 606228 MIM: 607355
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
AGO1 PubMed Links

Gene Details

Gene
AGO1
Gene Name
argonaute 1, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317122.1 1803 Intron NP_001304051.1
NM_001317123.1 1803 Intron NP_001304052.1
NM_012199.4 1803 Intron NP_036331.1
XM_011541236.2 1803 UTR 3 XP_011539538.1
XM_017001016.1 1803 UTR 3 XP_016856505.1
XM_017001017.1 1803 UTR 3 XP_016856506.1
XM_017001018.1 1803 UTR 3 XP_016856507.1
XM_017001019.1 1803 UTR 3 XP_016856508.1
Gene
AGO3
Gene Name
argonaute 3, RISC catalytic component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024852.3 1803 Intron NP_079128.2
NM_177422.2 1803 Intron NP_803171.1
XM_005270575.3 1803 UTR 5 XP_005270632.1
XM_005270576.3 1803 Intron XP_005270633.1
XM_011540879.2 1803 Intron XP_011539181.1
XM_011540882.2 1803 Intron XP_011539184.1
XM_017000523.1 1803 UTR 5 XP_016856012.1
XM_017000524.1 1803 Intron XP_016856013.1
XM_017000525.1 1803 Intron XP_016856014.1
XM_017000526.1 1803 Intron XP_016856015.1
XM_017000527.1 1803 Intron XP_016856016.1
XM_017000528.1 1803 Intron XP_016856017.1

View Full Product Details