Product Details

SNP ID
rs74988751
Assay Type
Functionally tested
NCBI dbSNP Submissions
25
Location
Chr.1:93869876 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTAATTGCGAAATTTCTTCTTCTTT[C/T]GGAACTTTTTTCCTGCTGCATTTGC
Phenotype
MIM: 611199
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DNTTIP2 PubMed Links

Gene Details

Gene
DNTTIP2
Gene Name
deoxynucleotidyltransferase terminal interacting protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014597.4 2304 Missense Mutation CAA,CGA Q749R NP_055412.2

View Full Product Details