Product Details

SNP ID
rs78459819
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:100539433 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCTACCAGGCGCTGTACCAGAGGC[G/T]CCACACGGCGGGCATGCTGGCGCTG
Phenotype
MIM: 607468
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GPR88 PubMed Links

Gene Details

Gene
GPR88
Gene Name
G protein-coupled receptor 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022049.2 871 Missense Mutation CGC,CTC R156L NP_071332.2

View Full Product Details