Product Details

SNP ID
rs78962608
Assay Type
Functionally tested
NCBI dbSNP Submissions
1
Location
Chr.1:100539440 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGGCGCTGTACCAGAGGCGCCACAC[A/G]GCGGGCATGCTGGCGCTGTCCTGGG
Phenotype
MIM: 607468
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GPR88 PubMed Links

Gene Details

Gene
GPR88
Gene Name
G protein-coupled receptor 88
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022049.2 878 Silent Mutation ACA,ACG T158T NP_071332.2

View Full Product Details