Product Details
- SNP ID
-
rs74626467
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
6
- Location
-
Chr.1:109750702 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAGCCTCCAGCCGGGACAGGATTCG[T/C]GGGGCCTCCTGTGGACATAGCATCT
- Phenotype
-
MIM: 614989
MIM: 138390
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
EPS8L3
PubMed Links
Gene Details
- Gene
- EPS8L3
- Gene Name
- EPS8 like 3
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001319952.1 |
1875 |
Silent Mutation |
CCA,CCG |
P513P |
NP_001306881.1 |
NM_024526.3 |
1875 |
Silent Mutation |
CCA,CCG |
P546P |
NP_078802.2 |
NM_133181.3 |
1875 |
Silent Mutation |
CCA,CCG |
P576P |
NP_573444.2 |
NM_139053.2 |
1875 |
Silent Mutation |
CCA,CCG |
P577P |
NP_620641.1 |
XM_011542132.1 |
1875 |
Silent Mutation |
CCA,CCG |
P585P |
XP_011540434.1 |
XM_011542133.1 |
1875 |
Silent Mutation |
CCA,CCG |
P584P |
XP_011540435.1 |
XM_011542134.2 |
1875 |
Silent Mutation |
CCA,CCG |
P551P |
XP_011540436.1 |
XM_011542135.2 |
1875 |
Silent Mutation |
CCA,CCG |
P521P |
XP_011540437.1 |
XM_017002327.1 |
1875 |
Intron |
|
|
XP_016857816.1 |
XM_017002328.1 |
1875 |
Silent Mutation |
CCA,CCG |
P555P |
XP_016857817.1 |
XM_017002329.1 |
1875 |
Silent Mutation |
CCA,CCG |
P547P |
XP_016857818.1 |
- Gene
- GSTM3
- Gene Name
- glutathione S-transferase mu 3
There are no transcripts associated with this gene.
View Full Product Details