Product Details

SNP ID
rs75048140
Assay Type
Functionally tested
NCBI dbSNP Submissions
11
Location
Chr.1:111499600 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAATGAGTCACCACCACACACA[C/T]GTTCAGCCCAACTGGAGCCTTTTGT
Phenotype
MIM: 600445
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ADORA3 PubMed Links

Gene Details

Gene
ADORA3
Gene Name
adenosine A3 receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000677.3 2074 UTR 3 NP_000668.1
NM_001302678.1 2074 UTR 3 NP_001289607.1
NM_001302679.1 2074 UTR 3 NP_001289608.1
Gene
TMIGD3
Gene Name
transmembrane and immunoglobulin domain containing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001081976.2 2074 Intron NP_001075445.1
NM_001302680.1 2074 Intron NP_001289609.1
NM_020683.6 2074 Intron NP_065734.5

View Full Product Details