Product Details

SNP ID
rs78581320
Assay Type
Functionally tested
NCBI dbSNP Submissions
16
Location
Chr.1:114394900 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GTAAATTGAAGTACAGCAAAAACTT[C/T]AAAAGCTTTTAAGAAAACATTATTA
Phenotype
MIM: 605769
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
TRIM33 PubMed Links

Gene Details

Gene
TRIM33
Gene Name
tripartite motif containing 33
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_015906.3 5491 UTR 3 NP_056990.3
NM_033020.2 5491 UTR 3 NP_148980.2
XM_005270936.3 5491 UTR 3 XP_005270993.1
XM_005270937.3 5491 UTR 3 XP_005270994.1
XM_011541568.2 5491 UTR 3 XP_011539870.1
XM_017001452.1 5491 UTR 3 XP_016856941.1
XM_017001453.1 5491 UTR 3 XP_016856942.1
XM_017001454.1 5491 Intron XP_016856943.1

View Full Product Details