Product Details
- SNP ID
-
rs77334515
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
14
- Location
-
Chr.1:117866433 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CAAAATGACTAAGGTAAGGTAAAGC[C/T]CTCCTTTCATTCTTCATGGTCTTTT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GDAP2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs150683842] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GDAP2
- Gene Name
- ganglioside induced differentiation associated protein 2
View Full Product Details