Product Details

SNP ID
rs78358435
Assay Type
Functionally tested
NCBI dbSNP Submissions
2
Location
Chr.1:38839689 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AATAGCTTTTCGGAAACAGTGGAAG[G/T]TGTAGTCTATTAAACCTGCAGGAAG
Phenotype
MIM: 608267
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RRAGC PubMed Links

Gene Details

Gene
RRAGC
Gene Name
Ras related GTP binding C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271851.1 1139 Missense Mutation AAC,ACC N321T NP_001258780.1
NM_022157.3 1139 Missense Mutation AAC,ACC N355T NP_071440.1

View Full Product Details