Product Details

SNP ID
rs11549874
Assay Type
Functionally tested
NCBI dbSNP Submissions
30
Location
Chr.1:38845945 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTTTTTAAAATGCTATTACCTTTTC[G/T]TTCAAAGCTTTCTTCCCTTAGAATG
Phenotype
MIM: 608267
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
RRAGC PubMed Links

Gene Details

Gene
RRAGC
Gene Name
Ras related GTP binding C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271851.1 1117 Silent Mutation AGA,CGA R314R NP_001258780.1
NM_022157.3 1117 Silent Mutation AGA,CGA R348R NP_071440.1

View Full Product Details