Product Details

SNP ID
rs76258765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:107327891 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTCGGTCTAGCTCCACCAGACTGT[A/G]TGGTGAGATCCTGCACAAACCACAT
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CWF19L2 PubMed Links

Gene Details

Gene
CWF19L2
Gene Name
CWF19-like 2, cell cycle control (S. pombe)
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_152434.2 Intron NP_689647.2
XM_011542620.2 Intron XP_011540922.1
XM_017017239.1 Intron XP_016872728.1
Gene
LOC105369477
Gene Name
uncharacterized LOC105369477
There are no transcripts associated with this gene.

View Full Product Details