Product Details

SNP ID
rs75640875
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:107505527 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTTCTAGGGCTATAAGCGGTAATA[C/T]GAAGCACTTTGACATTAATCATTTA
Phenotype
MIM: 613306
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
ALKBH8 PubMed Links

Gene Details

Gene
ALKBH8
Gene Name
alkB homolog 8, tRNA methyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301010.1 Intron NP_001287939.1
NM_138775.2 Intron NP_620130.2
XM_005271728.2 Intron XP_005271785.1
XM_005271729.3 Intron XP_005271786.1
XM_017018553.1 Intron XP_016874042.1
XM_017018554.1 Intron XP_016874043.1
XM_017018555.1 Intron XP_016874044.1
XM_017018556.1 Intron XP_016874045.1
XM_017018557.1 Intron XP_016874046.1

View Full Product Details