Product Details

SNP ID
rs80223723
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.11:113904870 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGTGGAGAGGAACCCTGTTAGGAG[A/G]AATTGAGCGGCATTCCATCTGGTAG
Phenotype
MIM: 604654
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
HTR3B PubMed Links
Additional Information
For this assay, SNP(s) [rs145648579] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
HTR3B
Gene Name
5-hydroxytryptamine receptor 3B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006028.4 75 UTR 5 NP_006019.1
XM_011543063.1 75 Intron XP_011541365.1
XM_017018552.1 75 Intron XP_016874041.1

View Full Product Details