Product Details

SNP ID
rs77529486
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:117215069 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCTCCCTCATAACCTCCTGTCAGC[A/G]GGGAGAGAGAAAAATCAGTCTTGCT
Phenotype
MIM: 604872
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PCSK7 PubMed Links

Gene Details

Gene
PCSK7
Gene Name
proprotein convertase subtilisin/kexin type 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004716.3 Intron NP_004707.2
XM_006718940.3 Intron XP_006719003.1
XM_017018546.1 Intron XP_016874035.1

View Full Product Details