Product Details

SNP ID
rs78276561
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:118163804 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCCACGGTAGGAAGAGGAAGAACTT[C/G]TCAGCATAAGTTAGGAAACAGGGTA
Phenotype
MIM: 601327
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SCN2B PubMed Links

Gene Details

Gene
SCN2B
Gene Name
sodium voltage-gated channel beta subunit 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_004588.4 3922 UTR 3 NP_004579.1

View Full Product Details