Product Details

SNP ID
rs79146737
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119007972 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTCAGGTCCTTGTTTCATACACCA[A/G]AAAGGACAACACTGAAACAAAAAGG
Phenotype
MIM: 180465
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC84 PubMed Links

Gene Details

Gene
CCDC84
Gene Name
coiled-coil domain containing 84
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198489.2 Intron NP_940891.1
XM_011542799.1 Intron XP_011541101.1
XM_011542800.2 Intron XP_011541102.1
XM_017017652.1 Intron XP_016873141.1
Gene
RPL23AP64
Gene Name
ribosomal protein L23a pseudogene 64
There are no transcripts associated with this gene.

Gene
RPS25
Gene Name
ribosomal protein S25
There are no transcripts associated with this gene.

View Full Product Details