Product Details

SNP ID
rs74992671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119096839 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGAGGGCAAGAAACGCCCAGGATG[C/T]CAATGATCACAGAGAAAGGAAAATG
Phenotype
MIM: 191350 MIM: 601772 MIM: 609806
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DPAGT1 PubMed Links

Gene Details

Gene
DPAGT1
Gene Name
dolichyl-phosphate N-acetylglucosaminephosphotransferase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001382.3 2024 UTR 3 NP_001373.2
XM_005271422.2 2024 UTR 3 XP_005271479.1
XM_011542648.1 2024 UTR 3 XP_011540950.1
XM_017017293.1 2024 UTR 3 XP_016872782.1
XM_017017294.1 2024 UTR 3 XP_016872783.1
XM_017017295.1 2024 UTR 3 XP_016872784.1
Gene
H2AFX
Gene Name
H2A histone family member X
There are no transcripts associated with this gene.

Gene
HMBS
Gene Name
hydroxymethylbilane synthase
There are no transcripts associated with this gene.

View Full Product Details