Product Details

SNP ID
rs78591480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:119152030 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACTTTTGTGTTAGGCATCAGTGTTC[A/C]ACCAGCTACTGTAATTAATTGAGGG
Phenotype
MIM: 607784
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
ABCG4 PubMed Links

Gene Details

Gene
ABCG4
Gene Name
ATP binding cassette subfamily G member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142505.1 Intron NP_001135977.1
NM_022169.4 Intron NP_071452.2
XM_011542952.2 Intron XP_011541254.1
XM_017018121.1 Intron XP_016873610.1

View Full Product Details