Product Details

SNP ID
rs77166726
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:123072548 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAAAGAAGAGACTATAATGGTATG[C/T]TGGGTAATCTTTCAGCATTCCTAGA
Phenotype
MIM: 611693
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CLMP PubMed Links

Gene Details

Gene
CLMP
Gene Name
CXADR like membrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024769.3 2413 UTR 3 NP_079045.1
XM_011542998.2 2413 Intron XP_011541300.1

View Full Product Details