Product Details

SNP ID
rs74498123
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:124959388 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGCCTAATATAATCTAATTAACTA[A/G]TGAAGACAGTAAATTTTTTTTGTAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC15 PubMed Links

Gene Details

Gene
CCDC15
Gene Name
coiled-coil domain containing 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_025004.2 Intron NP_079280.2
XM_017018346.1 Intron XP_016873835.1
XM_017018347.1 Intron XP_016873836.1
XM_017018348.1 Intron XP_016873837.1
XM_017018349.1 Intron XP_016873838.1
XM_017018350.1 Intron XP_016873839.1
XM_017018351.1 Intron XP_016873840.1
XM_017018352.1 Intron XP_016873841.1
XM_017018353.1 Intron XP_016873842.1

View Full Product Details