Product Details

SNP ID
rs78890235
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125577925 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TAAGCAGGTTTGTGTCAAGCAGCAC[A/G]TATCTATGGTTTCTTTGAATTGGAA
Phenotype
MIM: 605170
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
EI24 PubMed Links

Gene Details

Gene
EI24
Gene Name
EI24, autophagy associated transmembrane protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001290135.1 Intron NP_001277064.1
NM_004879.4 Intron NP_004870.3
XM_011543069.1 Intron XP_011541371.1
XM_011543070.1 Intron XP_011541372.1
XM_017018582.1 Intron XP_016874071.1
Gene
STT3A-AS1
Gene Name
STT3A antisense RNA 1
There are no transcripts associated with this gene.

View Full Product Details