Product Details

SNP ID
rs74527873
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:125957165 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTCCTCTCCTACCCACCTCCCTTCC[C/T]TCCAAGGAAACGCAAAGTTGCCCCG
Phenotype
MIM: 608707
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CDON PubMed Links

Gene Details

Gene
CDON
Gene Name
cell adhesion associated, oncogene regulated
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001243597.1 7730 UTR 3 NP_001230526.1
NM_016952.4 7730 UTR 3 NP_058648.4
XM_011542862.2 7730 UTR 3 XP_011541164.1
XM_011542863.2 7730 UTR 3 XP_011541165.1
XM_011542864.2 7730 UTR 3 XP_011541166.1
XM_011542865.2 7730 UTR 3 XP_011541167.1
XM_011542866.2 7730 UTR 3 XP_011541168.1
XM_017017873.1 7730 UTR 3 XP_016873362.1
Gene
LOC338667
Gene Name
V-set and immunoglobulin domain-containing protein 10-like
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_017018652.1 7730 Intron XP_016874141.1
XM_017018653.1 7730 Intron XP_016874142.1
XM_017018654.1 7730 Intron XP_016874143.1

View Full Product Details