Product Details

SNP ID
rs78558268
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:130090007 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCAGTAAATGAAATTTGAGGCCCT[A/G]TTCAACCTATAAGACCCAGTGGCTA
Phenotype
MIM: 104776
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
APLP2 PubMed Links

Gene Details

Gene
APLP2
Gene Name
amyloid beta precursor like protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001142276.1 Intron NP_001135748.1
NM_001142277.1 Intron NP_001135749.1
NM_001142278.1 Intron NP_001135750.1
NM_001243299.1 Intron NP_001230228.1
NM_001642.2 Intron NP_001633.1

View Full Product Details