Product Details

SNP ID
rs77684109
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:133840916 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ACCCCACTGTTCTCCGCGTTCCCAA[A/C]GCTCCATCTAGAGAGCAGAAACCCA
Phenotype
MIM: 609805
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
SPATA19 PubMed Links

Gene Details

Gene
SPATA19
Gene Name
spermatogenesis associated 19
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291992.1 613 Intron NP_001278921.1
NM_174927.2 613 UTR 3 NP_777587.1

View Full Product Details