Product Details

SNP ID
rs75335787
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.10:42796564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTTTCGGAGATGAAGATGAATCTGG[A/G]GATAGTGATGATGAAGAAGATGATG
Phenotype
MIM: 611448
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
BMS1 PubMed Links

Gene Details

Gene
BMS1
Gene Name
BMS1, ribosome biogenesis factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014753.3 1409 Silent Mutation GGA,GGG G440G NP_055568.3
XM_005271846.3 1409 Silent Mutation GGA,GGG G440G XP_005271903.1
XM_005271848.3 1409 Silent Mutation GGA,GGG G440G XP_005271905.1
XM_005271849.3 1409 Silent Mutation GGA,GGG G440G XP_005271906.1
XM_011540402.2 1409 Silent Mutation GGA,GGG G440G XP_011538704.1
XM_011540403.2 1409 Silent Mutation GGA,GGG G29G XP_011538705.1

View Full Product Details