Product Details

SNP ID
rs76193868
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:102373384 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCAGTTTCTTCTTTTTTCTTAACTT[C/T]TGGATTCTTCACAGTTACATCTGGG
Phenotype
MIM: 613365
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SLCO6A1 PubMed Links

Gene Details

Gene
SLCO6A1
Gene Name
solute carrier organic anion transporter family member 6A1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289002.1 1390 Missense Mutation AAA,GAA K710E NP_001275931.1
NM_001289004.1 1390 Missense Mutation AAA,GAA K648E NP_001275933.1
NM_001308014.1 1390 Missense Mutation AAA,GAA K457E NP_001294943.1
NM_173488.4 1390 Missense Mutation AAA,GAA K710E NP_775759.3
XM_005271874.3 1390 Missense Mutation AAA,GAA K710E XP_005271931.1
XM_011543147.2 1390 Missense Mutation AAA,GAA K675E XP_011541449.1
XM_011543148.2 1390 Missense Mutation AAA,GAA K631E XP_011541450.1
XM_011543150.1 1390 Missense Mutation AAA,GAA K467E XP_011541452.1
XM_011543153.2 1390 Missense Mutation AAA,GAA K436E XP_011541455.1
XM_011543154.2 1390 Intron XP_011541456.1

View Full Product Details