Product Details

SNP ID
rs77606373
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:108750674 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAAATAATTCAAGCTTTGGTAGACA[A/C]ATCATTTACATTTCATCTTCGATGT
Phenotype
MIM: 176942
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FER PubMed Links

Gene Details

Gene
FER
Gene Name
FER tyrosine kinase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308028.1 Intron NP_001294957.1
NM_001308031.1 Intron NP_001294960.1
NM_001308038.1 Intron NP_001294967.1
NM_005246.3 Intron NP_005237.2
XM_011543266.1 Intron XP_011541568.1
XM_011543267.1 Intron XP_011541569.1
XM_011543269.1 Intron XP_011541571.1
XM_011543270.2 Intron XP_011541572.1
XM_011543271.2 Intron XP_011541573.1
XM_011543272.1 Intron XP_011541574.1
XM_011543276.2 Intron XP_011541578.1
XM_011543278.1 Intron XP_011541580.1
XM_017009229.1 Intron XP_016864718.1
XM_017009230.1 Intron XP_016864719.1
XM_017009231.1 Intron XP_016864720.1
XM_017009232.1 Intron XP_016864721.1
XM_017009233.1 Intron XP_016864722.1
XM_017009234.1 Intron XP_016864723.1
XM_017009235.1 Intron XP_016864724.1

View Full Product Details