Product Details

SNP ID
rs78655000
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:127043583 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATTTCTAAATATCTTAATCTCCT[C/T]TGACCTTAAAAGAGTGAAATTGGCC
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C5orf63 PubMed Links

Gene Details

Gene
C5orf63
Gene Name
chromosome 5 open reading frame 63
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001164478.1 4655 Intron NP_001157950.1
NM_001164479.1 4655 UTR 3 NP_001157951.1
XM_017009464.1 4655 Intron XP_016864953.1
XM_017009465.1 4655 Intron XP_016864954.1

View Full Product Details