Product Details

SNP ID
rs77677325
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:134604590 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTAAACATGGCTTCCTTTCTAGA[C/G]GCTCGGGCCATAAAAATCTTATGTC
Phenotype
MIM: 607690
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
SAR1B PubMed Links

Gene Details

Gene
SAR1B
Gene Name
secretion associated Ras related GTPase 1B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001033503.2 3091 UTR 3 NP_001028675.1
NM_016103.3 3091 UTR 3 NP_057187.1
XM_017009521.1 3091 UTR 3 XP_016865010.1

View Full Product Details