Product Details

SNP ID
rs79805596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:135445507 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGCACTGCCCATGCCGGGCCTGAGG[A/G]GCCTGGCTCAGACTTTAGTATCCAA
Phenotype
MIM: 609710 MIM: 612663
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DCANP1 PubMed Links

Gene Details

Gene
DCANP1
Gene Name
dendritic cell associated nuclear protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_130848.2 1842 UTR 3 NP_570900.1
Gene
TIFAB
Gene Name
TIFA inhibitor
There are no transcripts associated with this gene.

View Full Product Details