Product Details

SNP ID
rs76327921
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138130419 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAAAAGTAACATGTAGTATAATGAT[A/G]CAGTAAAAAGTTTTTCATTTGAACG
Phenotype
MIM: 602848 MIM: 603575
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD8 PubMed Links

Gene Details

Gene
BRD8
Gene Name
bromodomain containing 8
There are no transcripts associated with this gene.

Gene
NME5
Gene Name
NME/NM23 family member 5
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003551.2 Intron NP_003542.1
XM_005272099.1 Intron XP_005272156.1
XM_006714716.3 Intron XP_006714779.1
XM_017009945.1 Intron XP_016865434.1

View Full Product Details