Product Details

SNP ID
rs78447328
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:132674351 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACTTGATTTGTTAACAGTTTTGTG[A/G]TAGGGAGAGTTGGAGGGAGGTTCAG
Phenotype
MIM: 616533 MIM: 604892
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DDX31 PubMed Links

Gene Details

Gene
DDX31
Gene Name
DEAD-box helicase 31
There are no transcripts associated with this gene.

Gene
GTF3C4
Gene Name
general transcription factor IIIC subunit 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012204.3 Intron NP_036336.2
XM_017015317.1 Intron XP_016870806.1
XM_017015318.1 Intron XP_016870807.1

View Full Product Details