Product Details

SNP ID
rs74427837
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:1290623 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACCTAGTTAGTAGAGGATTTGATA[C/T]GAAGCACCTGGGAAGGAATTCCTTA
Phenotype
MIM: 606801
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MAEA PubMed Links

Gene Details

Gene
MAEA
Gene Name
macrophage erythroblast attacher
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001017405.2 Intron NP_001017405.1
NM_001297430.1 Intron NP_001284359.1
NM_001297431.1 Intron NP_001284360.1
NM_001297432.1 Intron NP_001284361.1
NM_001297433.1 Intron NP_001284362.1
NM_005882.4 Intron NP_005873.2
XM_006713849.3 Intron XP_006713912.1
XM_006713850.2 Intron XP_006713913.1
XM_011513376.1 Intron XP_011511678.1
XM_011513377.1 Intron XP_011511679.1
XM_017007636.1 Intron XP_016863125.1
XM_017007637.1 Intron XP_016863126.1

View Full Product Details