Product Details

SNP ID
rs80148015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8322695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCAGAGTGAGCCCCTTCCTGCCAC[A/T]GTCACCCCAACTGAAATTGCCTTTC
Phenotype
MIM: 614611 MIM: 602139 MIM: 603685
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KANK3 PubMed Links

Gene Details

Gene
KANK3
Gene Name
KN motif and ankyrin repeat domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198471.2 2676 UTR 3 NP_940873.2
XM_006722718.3 2676 UTR 3 XP_006722781.1
XM_011527884.2 2676 UTR 3 XP_011526186.1
XM_017026563.1 2676 UTR 3 XP_016882052.1
Gene
NDUFA7
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
There are no transcripts associated with this gene.

Gene
RPS28
Gene Name
ribosomal protein S28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031.4 2676 Intron NP_001022.1

View Full Product Details