Product Details

SNP ID
rs79602012
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.19:8322790 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CTGTGCGCCAAAGGCTGAGGTGACT[A/G]ACGAGGAGATCTCCCCACAGCTAGG
Phenotype
MIM: 614611 MIM: 602139 MIM: 603685
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
KANK3 PubMed Links
Additional Information
For this assay, SNP(s) [rs4147653] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.

Gene Details

Gene
KANK3
Gene Name
KN motif and ankyrin repeat domains 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_198471.2 2581 UTR 3 NP_940873.2
XM_006722718.3 2581 UTR 3 XP_006722781.1
XM_011527884.2 2581 UTR 3 XP_011526186.1
XM_017026563.1 2581 UTR 3 XP_016882052.1
Gene
NDUFA7
Gene Name
NADH:ubiquinone oxidoreductase subunit A7
There are no transcripts associated with this gene.

Gene
RPS28
Gene Name
ribosomal protein S28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001031.4 2581 Intron NP_001022.1

View Full Product Details