Product Details
- SNP ID
-
hCV105188572
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.1:206009361 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AATAGAGAGGCACCAGCATTATCTG[T/C]CTCCTGACAGGATGCAACAAGAAGT
- Phenotype
-
MIM: 116890
- Polymorphism
- T/C, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CTSE
PubMed Links
Gene Details
- Gene
- CTSE
- Gene Name
- cathepsin E
There are no transcripts associated with this gene.
- Gene
- RAB7B
- Gene Name
- RAB7B, member RAS oncogene family
There are no transcripts associated with this gene.
View Full Product Details