Product Details

SNP ID
rs75904736
Assay Type
Functionally tested
NCBI dbSNP Submissions
12
Location
Chr.1:225402534 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TATACAATAGTGAATGAATTCTAAT[A/T]GTTCCAAAATTTTTAATTCACATTA
Phenotype
MIM: 603341 MIM: 600024
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DNAH14 PubMed Links

Gene Details

Gene
DNAH14
Gene Name
dynein axonemal heavy chain 14
There are no transcripts associated with this gene.

Gene
LBR
Gene Name
lamin B receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002296.3 2621 UTR 3 NP_002287.2
NM_194442.2 2621 UTR 3 NP_919424.1
XM_005273125.2 2621 UTR 3 XP_005273182.1
XM_011544185.2 2621 UTR 3 XP_011542487.1

View Full Product Details