Product Details
- SNP ID
-
rs75149861
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
19
- Location
-
Chr.1:227732652 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAGTGGTAAAACTCTTCAAAGTTGA[C/T]GCCCGAGCAGGACCTCATGATGACC
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
JMJD4
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs4837] are located under a probe and SNP(s) [rs751748] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- JMJD4
- Gene Name
- jumonji domain containing 4
- Gene
- SNAP47
- Gene Name
- synaptosome associated protein 47
View Full Product Details